Assay Details
Target Gene Details
Entrez Gene ID: | 68115 |
Gene Name: | RIKEN cDNA 9430016H08 gene |
Gene Aliases: |
AA409999, C78988 |
Location: |
Chr.1:57406548-57415958 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
9430016H08Rik | NM_001081181.2 | 4 | 946 | NP_001074650.1 |
AK007831.1 | 4 | 735 | ||
AK079116.1 | 4 | 948 | BAC37547.1 | |
BC024945.1 | 4 | 415 | AAH24945.1 |