Assay Details
Target Gene Details
Entrez Gene ID: | 329165 |
Gene Name: | abl-interactor 2 |
Gene Aliases: |
8430425M24Rik, AI839867, C130078H13 |
Location: |
Chr.1:60408988-60481945 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abi2 | NM_001198570.1 | 9 | 1295 | NP_001185499.1 |
NM_001198571.1 | 9 | 1196 | NP_001185500.1 | |
NM_198127.2 | 7 | 1094 | NP_937760.1 | |
BC056345.1 | 7 | 915 | AAH56345.1 | |
BC079545.1 | 9 | 1081 | AAH79545.1 | |
BC079646.1 | 9 | 1219 | AAH79646.1 |
More Information
Set Membership: |
Intragenic Exonic |