Assay Details
Target Gene Details
Entrez Gene ID: | 67198 |
Gene Name: | spermatogenesis associated, serine-rich 2-like |
Gene Aliases: |
2810022L02Rik, A230104H11Rik, AA987173, AI480531, AI842453, AW413586 |
Location: |
Chr.1:57774162-57948398 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spats2l | NM_001164566.1 | NP_001158038.1 | ||
NM_144882.4 | NP_659131.2 | |||
AK012791.1 | ||||
AK019292.1 | ||||
AK031266.1 | BAC27328.1 | |||
AK032169.1 | BAC27738.1 | |||
BC014764.1 | AAH14764.1 | |||
BC021879.1 | AAH21879.1 | |||
BY069430.1 | ||||
BY739337.1 |