Assay Details
Target Gene Details
Entrez Gene ID: | 215243 |
Gene Name: | TRAF3 interacting protein 3 |
Gene Aliases: |
6030423D04Rik, AI115021, T3jam |
Location: |
Chr.1:193175504-193201546 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Traf3ip3 | NM_153137.4 | NP_694777.3 | ||
AK031400.1 | BAC27388.1 | |||
AK036608.1 | ||||
AK161211.1 | ||||
AY383616.1 | AAQ84304.1 | |||
BC137605.1 | ||||
BC171933.1 |
More Information
Set Membership: |
Intragenic Non-exonic |