Assay Details
Target Gene Details
Entrez Gene ID: | 68691 |
Gene Name: | KAT8 regulatory NSL complex subunit 1-like |
Gene Aliases: |
1110028C15Rik, 1190030G24, AI427598, C430010P07Rik, mKIAA4189 |
Location: |
Chr.1:66719243-66817596 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kansl1l | NM_001122738.1 | 14 | 2776 | NP_001116210.1 |
NM_177645.4 | 13 | 2749 | NP_808313.3 | |
AK082897.1 | 13 | 1596 | BAC38675.1 | |
AK220534.1 | 13 | 2700 | BAD90312.1 | |
BC141036.1 | 14 | 2659 |