Assay Details
Target Gene Details
Entrez Gene ID: | 69171 |
Gene Name: | cyclin Pas1/PHO80 domain containing 1 |
Gene Aliases: |
1810031K17Rik, AU043960 |
Location: |
Chr.1:75135215-75142733 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cnppd1 | NM_026977.2 | NP_081253.1 | ||
AK029090.1 | BAC26289.1 | |||
AK145962.1 | BAE26787.1 | |||
AK169779.1 | BAE41361.1 | |||
AK177396.1 | ||||
AK177413.1 | ||||
AK204709.1 | ||||
BC013714.1 | ||||
BC028815.1 | AAH28815.1 |