Assay Details
Target Gene Details
Entrez Gene ID: | 98685 |
Gene Name: | tRNA methyltransferase 1 like |
Gene Aliases: |
1190005F20Rik, AW226554, C1orf25, Trm1-like, Trm1l |
Location: |
Chr.1:151428547-151458183 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trmt1l | NM_026876.3 | 12 | 1933 | NP_081152.2 |
AK004493.1 | 4 | 426 | BAB23331.1 | |
AK146088.1 | 12 | 1934 | BAE26891.1 | |
BC022682.1 | 7 | 969 | AAH22682.1 | |
BC037478.1 | 8 | 1113 | AAH37478.1 | |
BC110425.1 | 12 | 1916 | ||
BC132299.1 | 12 | 1791 |
More Information
Set Membership: |
Intragenic Exonic |