Assay Details
Target Gene Details
Entrez Gene ID: | 66052 |
Gene Name: | succinate dehydrogenase complex, subunit C, integral membrane protein |
Gene Aliases: |
0610010E03Rik, AI316496, AU019277 |
Location: |
Chr.1:171129157-171150603 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sdhc | NM_025321.3 | NP_079597.2 | ||
AK002459.1 | BAB22116.1 | |||
AK012818.1 | BAB28491.1 | |||
AK135503.1 | BAE22557.1 | |||
BC005779.1 | AAH05779.1 | |||
BC083091.1 | AAH83091.1 | |||
BY711932.1 |
More Information
Set Membership: |
Intragenic Non-exonic |