Assay Details
Target Gene Details
Entrez Gene ID: | 78825 |
Gene Name: | desumoylating isopeptidase 2 |
Gene Aliases: |
5830417C01Rik, DeSI-2, Fam152a, Pppde1 |
Location: |
Chr.1:178187231-178252598 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Desi2 | NM_024282.3 | NP_077244.1 | ||
AK020014.1 | BAB31967.1 | |||
AK029574.1 | BAC26520.1 | |||
AK049579.1 | BAC33822.1 | |||
AK146492.1 | BAE27211.1 | |||
AK160733.1 | BAE35977.1 | |||
AK216786.1 | ||||
BC002200.1 | AAH02200.1 | |||
BC046816.1 | AAH46816.1 |
More Information
Set Membership: |
Intragenic Non-exonic |