Assay Details
Target Gene Details
Entrez Gene ID: | 27392 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class N |
Gene Aliases: |
Gm20308, PIG-N |
Location: |
Chr.1:105518422-105663722 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pign | NM_013784.3 | 25 | 2732 | NP_038812.2 |
AB030279.1 | 25 | 2718 | BAA82619.1 | |
AB030280.1 | 25 | 2718 | BAA82620.1 | |
AK132928.1 | 4 | 647 | BAE21427.1 | |
BC021148.1 | 25 | 2716 | AAH21148.1 |
More Information
Set Membership: |
Intragenic Exonic |