Assay Details
Target Gene Details
Entrez Gene ID: | 226856 |
Gene Name: | lysophosphatidylglycerol acyltransferase 1 |
Gene Aliases: |
AI649174, AW112037, BC013667 |
Location: |
Chr.1:191717556-191784255 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lpgat1 | NM_001134829.1 | NP_001128301.1 | ||
NM_172266.3 | NP_758470.1 | |||
AK133814.1 | BAE21857.1 | |||
AK172914.1 | BAD32192.1 | |||
BC013667.1 | AAH13667.1 | |||
BC024750.1 | AAH24750.1 |
More Information
Set Membership: |
Intragenic Non-exonic |