Assay Details
Target Gene Details
Entrez Gene ID: | 18777 |
Gene Name: | lysophospholipase 1 |
Gene Aliases: |
Pla1a |
Location: |
Chr.1:4807563-4846737 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lypla1 | NM_008866.2 | NP_032892.1 | ||
AK002674.1 | BAB22276.1 | |||
AK050549.1 | BAC34318.1 | |||
AK146874.1 | BAE27497.1 | |||
BC013536.1 | AAH13536.1 | |||
BC052848.1 | 1 | 43 | AAH52848.1 | |
CT010201.1 | CAJ18409.1 | |||
U89352.1 | AAB48627.1 |
More Information
Set Membership: |
Intragenic Non-exonic |