Assay Details
Target Gene Details
Entrez Gene ID: | 55927 |
Gene Name: | hairy and enhancer of split 6 |
Gene Aliases: |
AI326893, bHLHb41 |
Location: |
Chr.1:91411482-91413877 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hes6 | NM_019479.3 | 2 | 211 | NP_062352.1 |
AB035178.1 | 2 | 179 | BAA96081.1 | |
AF247040.1 | 2 | 126 | AAF63757.1 | |
AF260236.1 | 2 | 183 | AAK51633.1 | |
AK005022.1 | 2 | 214 | ||
AK007480.1 | 2 | 220 | ||
AK012582.1 | 2 | 217 | ||
AK013443.1 | 2 | 206 | BAB28856.1 | |
AK076135.1 | 2 | 211 | BAC36210.1 | |
AK146192.1 | 2 | 178 | ||
AK155340.1 | 2 | 212 | BAE33204.1 | |
BC012897.1 | 2 | 189 | AAH12897.1 |
More Information
Set Membership: |
Intragenic Exonic |