Assay Details
Target Gene Details
Entrez Gene ID: | 269109 |
Gene Name: | dipeptidylpeptidase 10 |
Gene Aliases: |
6430601K09Rik, DPP X, Dprp3 |
Location: |
Chr.1:123332138-124845960 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dpp10 | NM_199021.3 | NP_950186.3 | ||
AK032579.1 | ||||
AK046842.1 | ||||
BC067026.1 | AAH67026.1 |
More Information
Set Membership: |
Intragenic Non-exonic |