Assay Details
Target Gene Details
Entrez Gene ID: | 12902 |
Gene Name: | complement receptor 2 |
Gene Aliases: |
C3DR, CD21, CD35, Cr-1, Cr-2, Cr1 |
Location: |
Chr.1:195136811-195177190 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cr2 | NM_007758.2 | NP_031784.1 | ||
AK003072.2 | BAB22548.1 | |||
AK007487.1 | BAB25063.1 | |||
BC109344.1 | AAI09345.1 | |||
BC109345.1 | AAI09346.1 | |||
M29281.1 | AAA37447.1 | |||
M35684.1 | AAA37448.1 | |||
M61132.1 | AAA63295.1 |