Assay Details
Target Gene Details
Entrez Gene ID: | 213043 |
Gene Name: | aldehyde oxidase 2 |
Gene Aliases: |
Aoh3, Aox3l1 |
Location: |
Chr.1:58276141-58379264 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aox2 | NM_001008419.2 | 7 | 648 | NP_001008419.1 |
AY187018.2 | 7 | 648 | AAO38750.2 | |
AY665589.1 | 7 | 555 | AAV68256.1 | |
BC150826.1 | 7 | 575 |