Assay Details
Target Gene Details
Entrez Gene ID: | 67198 |
Gene Name: | spermatogenesis associated, serine-rich 2-like |
Gene Aliases: |
2810022L02Rik, A230104H11Rik, AA987173, AI480531, AI842453, AW413586 |
Location: |
Chr.1:57774162-57948398 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spats2l | NM_144882.4 | 1 | 74 | NP_659131.2 |
AK012791.1 | ||||
AK019292.1 | ||||
AK020712.1 | ||||
AK031266.1 | BAC27328.1 | |||
AK032169.1 | BAC27738.1 | |||
AK161988.1 | ||||
BC021879.1 | AAH21879.1 | |||
BY317793.1 | 1 | 67 |