Assay Details
Target Gene Details
Entrez Gene ID: | 75472 |
Gene Name: | cilia and flagella associated protein 126 |
Gene Aliases: |
1700009P17Rik, AV282292, Flattop, Fltp |
Location: |
Chr.1:171113744-171126967 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cfap126 | NM_001081275.1 | NP_001074744.1 | ||
AK005804.1 | BAB24245.1 | |||
BC061017.1 | AAH61017.1 |