Assay Details
Target Gene Details
Entrez Gene ID: | 18189 |
Gene Name: | neurexin I |
Gene Aliases: |
1700062G21Rik, 9330127H16Rik, A230068P09Rik, mKIAA0578 |
Location: |
Chr.17:90033644-91093100 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrxn1 | NM_020252.3 | 23 | 8885 | NP_064648.3 |
NM_177284.2 | 21 | 8849 | NP_796258.2 | |
AK045351.1 | 1 | 2667 |