Assay Details
Target Gene Details
Entrez Gene ID: | 18189 |
Gene Name: | neurexin I |
Gene Aliases: |
1700062G21Rik, 9330127H16Rik, A230068P09Rik, mKIAA0578 |
Location: |
Chr.17:90033644-91093100 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrxn1 | NM_020252.3 | 6 | 2079 | NP_064648.3 |
NM_177284.2 | 5 | 2067 | NP_796258.2 | |
AB093249.2 | 6 | 2079 | BAC41433.2 | |
AK037724.1 | 3 | 2010 | ||
AK133271.1 | 2 | 694 | ||
AK137407.1 | 6 | 2086 | ||
AK163484.1 | 3 | 1989 |