Assay Details
Target Gene Details
Entrez Gene ID: | 18736 |
Gene Name: | POU domain, class 1, transcription factor 1 |
Gene Aliases: |
GHF-1, Hmp1, Pit-1, Pit1, Pit1-rs1, dw, dwarf |
Location: |
Chr.16:65520529-65534146 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pou1f1 | NM_008849.4 | NP_032875.1 | ||
AK133615.1 | ||||
X57512.1 | CAA40737.1 |