Assay Details
Target Gene Details
Entrez Gene ID: | 121022 |
Gene Name: | mitochondrial ribosomal protein S6 |
Gene Aliases: |
AW046321 |
Location: |
Chr.16:92058336-92112227 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mrps6 | NM_080456.1 | 2 | 220 | NP_536704.1 |
AB049943.1 | 2 | 235 | BAB40996.1 | |
AK165590.1 | 2 | 237 | BAE38278.1 | |
AK166374.1 | 2 | 235 | BAE38738.1 | |
AK167304.1 | 2 | 235 | BAE39408.1 | |
AK167316.1 | 2 | 235 | BAE39418.1 | |
AY061856.1 | 2 | 235 | AAL35740.1 | |
BC027271.1 | 2 | 220 | AAH27271.1 |
More Information
Set Membership: |
Intragenic Exonic |