Assay Details
Target Gene Details
Entrez Gene ID: | 20658 |
Gene Name: | Son DNA binding protein |
Gene Aliases: |
2900011L12Rik, AA409051, AU067731, C81487, mKIAA1019, nrebp |
Location: |
Chr.16:91647824-91679195 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Son | NM_178880.4 | 6 | 6641 | NP_849211.3 |
AB546195.1 | 7 | 6456 | ||
AK122418.1 | 4 | 1002 | BAC65700.1 | |
DQ549242.1 |
More Information
Set Membership: |
Intragenic Exonic |