Assay Details
Target Gene Details
Entrez Gene ID: | 28080 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit |
Gene Aliases: |
ATPO, D12Wsu28e, OSCP |
Location: |
Chr.16:91925223-91931630 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp5o | NM_138597.2 | NP_613063.1 | ||
AK005309.1 | BAB23945.1 | |||
AK168292.1 | BAE40236.1 | |||
AK169318.1 | BAE41073.1 | |||
BC012241.1 | AAH12241.1 |
More Information
Set Membership: |
Intragenic Non-exonic |