Assay Details
Target Gene Details
Entrez Gene ID: | 14450 |
Gene Name: | phosphoribosylglycinamide formyltransferase |
Gene Aliases: |
Gaps, Prgs |
Location: |
Chr.16:91621395-91647481 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gart | NM_010256.2 | 4 | 505 | NP_034386.2 |
AK051812.1 | 1 | 3381 | ||
AK146355.1 | 4 | 490 | BAE27107.1 | |
AK146478.1 | 4 | 443 | ||
AK168445.1 | 4 | 455 | BAE40349.1 | |
AK168458.1 | 4 | 465 | ||
AK168501.1 | 4 | 468 | BAE40386.1 | |
AK168724.1 | 4 | 480 | BAE40565.1 | |
AK168796.1 | 4 | 480 | BAE40629.1 | |
AK168864.1 | 4 | 456 | BAE40683.1 | |
AK168876.1 | 4 | 457 | BAE40694.1 | |
AK177198.1 | ||||
AK183808.1 | ||||
AK183944.1 | ||||
AK204723.1 | ||||
AK205057.1 | ||||
BC070465.1 | 4 | 452 | AAH70465.1 | |
U01023.1 | 4 | 476 | AAA19012.1 | |
U01024.1 | 3 | 410 | AAA19013.1 |