Assay Details
Target Gene Details
Entrez Gene ID: | 14450 |
Gene Name: | phosphoribosylglycinamide formyltransferase |
Gene Aliases: |
Gaps, Prgs |
Location: |
Chr.16:91621395-91647481 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gart | NM_010256.2 | 2 | 161 | NP_034386.2 |
AK051812.1 | 1 | 1563 | ||
AK146355.1 | 2 | 146 | BAE27107.1 | |
AK146478.1 | 2 | 98 | ||
AK168445.1 | 2 | 111 | BAE40349.1 | |
AK168458.1 | 2 | 121 | ||
AK168501.1 | 2 | 124 | BAE40386.1 | |
AK168724.1 | 2 | 136 | BAE40565.1 | |
AK168796.1 | 2 | 136 | BAE40629.1 | |
AK168864.1 | 2 | 112 | BAE40683.1 | |
AK168876.1 | 2 | 113 | BAE40694.1 | |
AK177198.1 | ||||
AK183944.1 | ||||
AK204723.1 | ||||
AK205057.1 | ||||
BB843046.1 | 2 | 162 | ||
BC070465.1 | 2 | 108 | AAH70465.1 | |
U01023.1 | 2 | 132 | AAA19012.1 | |
U01024.1 | 1 | 66 | AAA19013.1 |