Assay Details
Target Gene Details
Entrez Gene ID: | 14450 |
Gene Name: | phosphoribosylglycinamide formyltransferase |
Gene Aliases: |
Gaps, Prgs |
Location: |
Chr.16:91621395-91647481 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gart | NM_010256.2 | NP_034386.2 | ||
AK146355.1 | BAE27107.1 | |||
AK146478.1 | ||||
AK168458.1 | ||||
AK168501.1 | BAE40386.1 | |||
AK168724.1 | BAE40565.1 | |||
AK168796.1 | BAE40629.1 | |||
AK168864.1 | BAE40683.1 | |||
AK168876.1 | BAE40694.1 | |||
AK181084.1 | ||||
AK184743.1 | ||||
AK212300.1 | ||||
BC070465.1 | AAH70465.1 | |||
U01024.1 | AAA19013.1 |