Assay Details
Target Gene Details
Entrez Gene ID: | 56088 |
Gene Name: | proteasome (prosome, macropain) assembly chaperone 1 |
Gene Aliases: |
AW552102, Dscr2 |
Location: |
Chr.16:95979935-95990903 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psmg1 | NM_019537.2 | 5 | 543 | NP_062410.1 |
AJ238270.1 | 5 | 545 | CAB87195.1 | |
AK042203.1 | 5 | 554 | BAC31197.1 | |
AK088118.1 | 5 | 556 | BAC40157.1 | |
BC032965.1 | 5 | 536 | AAH32965.1 | |
BC060285.1 | 5 | 557 | AAH60285.1 |
More Information
Set Membership: |
Intragenic Exonic |