Assay Details
Target Gene Details
Entrez Gene ID: | 22129 |
Gene Name: | tetratricopeptide repeat domain 3 |
Gene Aliases: |
2610202A04Rik, AA409221, D16Ium21, D16Ium21e, TPRD, mKIAA4119 |
Location: |
Chr.16:94370739-94469222 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc3 | NM_009441.2 | NP_033467.2 | ||
AB008516.1 | BAA31563.1 | |||
AK053765.1 | BAC35513.1 | |||
AK084201.1 | ||||
AK088273.1 | BAC40250.1 | |||
AK165850.1 | BAE38412.1 | |||
AK166120.1 | ||||
AK166164.1 | ||||
AK220495.1 | BAD90293.1 | |||
BC019173.1 | ||||
BC057207.1 | AAH57207.1 |
More Information
Set Membership: |
Intragenic Non-exonic |