Assay Details
Target Gene Details
Entrez Gene ID: | 60364 |
Gene Name: | downstream neighbor of SON |
Gene Aliases: |
1110025J21Rik, AI845729, ORF60 |
Location: |
Chr.16:91666856-91688759 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Donson | NM_021720.1 | 10 | 2078 | NP_068366.1 |
AF193608.1 | 7 | 1369 | AAF23122.1 | |
AK042415.1 | 9 | 2225 | ||
AK132269.1 | 10 | 1957 | ||
BC043316.1 | 10 | 2078 | AAH43316.1 |