Assay Details
Target Gene Details
Entrez Gene ID: | 15980 |
Gene Name: | interferon gamma receptor 2 |
Gene Aliases: |
Ifgr2, Ifgt |
Location: |
Chr.16:91547094-91564007 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ifngr2 | NM_008338.3 | 7 | 1383 | NP_032364.1 |
AK165193.1 | 7 | 1399 | BAE38071.1 | |
AK169907.1 | 7 | 1404 | ||
BC055745.1 | 7 | 1383 | AAH55745.1 |
More Information
Set Membership: |
Intragenic Exonic |