Assay Details
Target Gene Details
Entrez Gene ID: | 71446 |
Gene Name: | tryptophan rich basic protein |
Gene Aliases: |
5530402J05Rik, C030018G21Rik, Chd5 |
Location: |
Chr.16:96145354-96189247 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wrb | NM_207301.2 | 5 | 2062 | NP_997184.1 |
AK017448.1 | 1 | 1205 | ||
BC031769.1 | 5 | 2062 | AAH31769.1 |