Assay Details
Target Gene Details
Entrez Gene ID: | 66609 |
Gene Name: | crystallin, zeta (quinone reductase)-like 1 |
Gene Aliases: |
2210407J23Rik, 2410006O11Rik, QOH-1 |
Location: |
Chr.16:91689322-91728802 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cryzl1 | NM_026994.1 | 12 | 1239 | NP_081270.1 |
NM_133679.2 | 13 | 1284 | NP_598440.1 | |
AK008846.1 | 13 | 1057 | ||
AK017995.1 | 12 | 1232 | ||
AK145827.1 | 13 | 1108 | BAE26679.1 | |
AK170997.1 | 5 | 399 | ||
BC010479.1 | 13 | 1081 | AAH10479.1 | |
BC019387.1 | 12 | 1048 | AAH19387.1 | |
BU757748.1 |
Target Gene Details
Entrez Gene ID: | 100038392 |
Gene Name: | predicted gene 10785 |
Gene Aliases: |
ENSMUSG00000074911 |
Location: |
Chr.16:91688898-91715755 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm10785 | NR_040389.1 | |||
AK038992.1 | ||||
AK160609.1 | BAE35912.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |