Assay Details
Target Gene Details
Entrez Gene ID: | 16183 |
Gene Name: | interleukin 2 |
Gene Aliases: |
Il-2 |
Location: |
Chr.3:37120713-37125954 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il2 | NM_008366.3 | 1 | 59 | NP_032392.1 |
AF065914.1 | 1 | 57 | AAD25890.1 | |
AF065915.1 | 1 | 57 | AAD25891.1 | |
AF065916.1 | 1 | 57 | AAD25892.1 | |
AF538059.1 | 1 | 64 | AAN77291.1 | |
AF542383.1 | 1 | 64 | AAN77288.1 | |
AF542384.1 | 1 | 64 | AAN77289.1 | |
AF542385.1 | 1 | 64 | AAN77290.1 | |
AY147902.1 | AAN38301.1 | |||
BC116845.1 | 1 | 57 | ||
BC116873.1 | 1 | 57 | ||
K02292.1 | 1 | 59 | AAA39289.1 | |
U41505.1 | AAB39209.1 | |||
U41506.1 | AAB39210.1 | |||
X01772.1 | 1 | 58 | CAA25909.1 | |
X66058.1 | CAA46854.1 | |||
X73040.1 | 1 | 58 |
More Information
Set Membership: |
Intragenic Exonic |