Assay Details
Target Gene Details
Entrez Gene ID: | 55858 |
Gene Name: | transmembrane protein 165 |
Gene Aliases: |
CDG2K, FT27, GDT1, TMPT27, TPARL |
Location: |
Chr.4:55395913-55453397 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM165 | NM_018475.4 | NP_060945.2 | ||
NR_073070.1 | ||||
XM_011534394.2 | XP_011532696.1 | |||
XM_017008412.1 | XP_016863901.1 | |||
AF132746.1 | AAL75947.1 | |||
AF183409.1 | AAG09678.1 | |||
AF220188.1 | AAF67653.1 | |||
AK290663.1 | ||||
AK295289.1 | ||||
BC104978.1 | AAI04979.1 | |||
BC104980.1 | AAI04981.1 | |||
BC107582.1 | AAI07583.1 | |||
DA371259.1 | ||||
HY089602.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2667161 | Chr.4:55400600 - 55402953 on Build GRCh38 | Deletion |
![]() |
esv2660719 | Chr.4:55399828 - 55403725 on Build GRCh38 | Deletion |
![]() |
esv3600549 | Chr.4:55400563 - 55402854 on Build GRCh38 | Loss |
![]() |
esv23054 | Chr.4:55399954 - 55401777 on Build GRCh38 | Loss |
![]() |
nsv594204 | Chr.4:55400806 - 55402340 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)