Assay Details
Target Gene Details
Entrez Gene ID: | 65009 |
Gene Name: | NDRG family member 4 |
Gene Aliases: |
BDM1, SMAP-8, SMAP8 |
Location: |
Chr.16:58463645-58515403 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDRG4 | NM_001130487.1 | NP_001123959.1 | ||
NM_020465.3 | NP_065198.1 | |||
XM_006721256.3 | XP_006721319.1 | |||
XM_006721258.3 | XP_006721321.1 | |||
XM_006721260.3 | XP_006721323.1 | |||
XM_011523294.2 | XP_011521596.1 | |||
XM_017023587.1 | XP_016879076.1 | |||
XM_017023588.1 | XP_016879077.1 | |||
XM_017023590.1 | XP_016879079.1 | |||
AB021172.1 | BAB20288.1 | |||
CF552009.1 | ||||
DA242460.1 | ||||
DB498690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv25662 | Chr.16:58463738 - 58464523 on Build GRCh38 | Loss | NDRG4 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |