Assay Details
Target Gene Details
Entrez Gene ID: | 84690 |
Gene Name: | spermatogenesis associated 22 |
Gene Aliases: |
NYD-SP20, NYDSP20 |
Location: |
Chr.17:3440011-3513852 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPATA22 | NM_001321336.1 | NP_001308265.1 | ||
NM_001321337.1 | NP_001308266.1 | |||
AK295299.1 |
Target Gene Details
Entrez Gene ID: | 162514 |
Gene Name: | transient receptor potential cation channel subfamily V member 3 |
Gene Aliases: |
FNEPPK2, OLMS, VRL3 |
Location: |
Chr.17:3510502-3557995 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TRPV3 | NM_001258205.1 | 18 | 4947 | NP_001245134.1 |
NM_145068.3 | 18 | 4944 | NP_659505.1 | |
AK127330.1 | 6 | 2943 | ||
AK127726.1 | 12 | 3896 | ||
BX537539.1 | 12 | 3884 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892950 | Chr.17:3131812 - 3577153 on Build GRCh38 | Loss | OR3A4P OR3A1 OR1D4 ASPA OR3A3 OR3A2 OR1A1 SPATA22 OR1E1 TRPV1 OR1E2 TRPV3 OR1A2 |
nsv574237 | Chr.17:3486822 - 3729542 on Build GRCh38 | Loss | SHPK EMC6 P2RX5-TAX1BP3 ITGAE TAX1BP3 SPATA22 GSG2 P2RX5 ASPA TRPV1 TRPV3 CTNS |
nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain | OR3A4P NCBP3 ATP2A3 OR1D5 ITGAE MIR6776 ASPA OR3A2 SPATA22 P2RX5 OR1D2 OR1E1 OR1E2 CCDC92B P2RX1 OR1A2 LOC105371591 SHPK MIR1253 OR1G1 OR3A1 OR1D4 CLUH LOC105371592 OR3A3 LOC101927911 EMC6 P2RX5-TAX1BP3 CAMKK1 RAP1GAP2 TAX1BP3 OR1A1 GSG2 TRPV1 TRPV3 CTNS PAFAH1B1 |
esv23898 | Chr.17:3511464 - 3512895 on Build GRCh38 | Gain | SPATA22 TRPV3 |
esv2660206 | Chr.17:2679367 - 3519614 on Build GRCh38 | Deletion | OR3A4P MIR1253 OR1D5 OR1G1 OR3A1 MIR6776 OR1D4 ASPA CLUH LOC105371592 OR3A3 LOC101927911 RAP1GAP2 OR3A2 OR1A1 SPATA22 OR1D2 OR1E1 OR1E2 CCDC92B TRPV3 OR1A2 PAFAH1B1 LOC105371591 |
nsv1055740 | Chr.17:3381841 - 3890153 on Build GRCh38 | Gain | NCBP3 SHPK ITGAE ASPA OR3A3 EMC6 P2RX5-TAX1BP3 CAMKK1 OR3A2 TAX1BP3 SPATA22 GSG2 P2RX5 OR1E1 TRPV1 OR1E2 TRPV3 CTNS |
nsv833341 | Chr.17:3352243 - 3521390 on Build GRCh38 | Loss | OR3A2 SPATA22 ASPA OR1E1 OR1E2 TRPV3 OR3A3 |
nsv1060071 | Chr.17:3398474 - 3633472 on Build GRCh38 | Loss | SHPK SPATA22 ASPA TRPV1 OR1E2 TRPV3 OR3A3 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |