Assay Details
Target Gene Details
Entrez Gene ID: | 23417 |
Gene Name: | malonyl-CoA decarboxylase |
Gene Aliases: |
MCD |
Location: |
Chr.16:83899125-83916182 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MLYCD | NM_012213.2 | NP_036345.2 | ||
AF090834.1 | AAD48994.1 | |||
AF097832.2 | AAD16177.2 | |||
AF153679.1 | AAD34631.1 | |||
BC000286.1 | AAH00286.1 | |||
BC052592.1 | AAH52592.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1908 | Chr.16:83876721 - 83909333 on Build GRCh38 | Insertion |
![]() |
nsv573401 | Chr.16:83679394 - 83940461 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
dgv3054n100 | Chr.16:83519838 - 83940461 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv573398 | Chr.16:83516215 - 83940461 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv573408 | Chr.16:83902155 - 83902816 on Build GRCh38 | Gain |
![]() |
esv26248 | Chr.16:83901927 - 83902935 on Build GRCh38 | Loss |
![]() |
nsv457587 | Chr.16:83817468 - 83908216 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)