Assay Details
Target Gene Details
Entrez Gene ID: | 11217 |
Gene Name: | A-kinase anchoring protein 2 |
Gene Aliases: |
AKAP-2, AKAPKL, MISP2, PRKA2 |
Location: |
Chr.9:110048598-110172512 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AKAP2 | NM_001004065.4 | NP_001004065.2 | ||
NM_001198656.1 | NP_001185585.1 | |||
AK057098.1 | ||||
AK304311.1 | ||||
BC146863.1 | ||||
BC171800.1 | ||||
BE674805.1 |
Target Gene Details
Entrez Gene ID: | 445815 |
Gene Name: | PALM2-AKAP2 readthrough |
Gene Aliases: |
AKAP2 |
Location: |
Chr.9:109780297-110172512 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PALM2-AKAP2 | NM_007203.4 | NP_009134.1 | ||
NM_147150.2 | NP_671492.1 | |||
AB023137.1 | ||||
AJ303079.1 | CAC38839.1 | |||
AJ312217.1 | CAC59702.1 | |||
BC140818.1 | ||||
BX649126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv494n21 | Chr.9:109952819 - 110202456 on Build GRCh38 | Gain | AKAP2 PALM2-AKAP2 C9orf152 |
esv3573350 | Chr.9:110061538 - 110069597 on Build GRCh38 | Loss | AKAP2 PALM2-AKAP2 |
nsv1150859 | Chr.9:110067907 - 110068654 on Build GRCh38 | Deletion | AKAP2 PALM2-AKAP2 |
nsv466480 | Chr.9:110067939 - 110096621 on Build GRCh38 | Loss | AKAP2 PALM2-AKAP2 |
esv28884 | Chr.9:110067919 - 110068852 on Build GRCh38 | Loss | AKAP2 PALM2-AKAP2 |
esv3621448 | Chr.9:109954078 - 110129324 on Build GRCh38 | Gain | AKAP2 PALM2-AKAP2 |
esv3621452 | Chr.9:110067914 - 110068620 on Build GRCh38 | Loss | AKAP2 PALM2-AKAP2 |
nsv1043494 | Chr.9:109988131 - 110214627 on Build GRCh38 | Gain | AKAP2 PALM2-AKAP2 C9orf152 |
nsv1117903 | Chr.9:110067915 - 110068618 on Build GRCh38 | Deletion | AKAP2 PALM2-AKAP2 |
esv3621449 | Chr.9:109959589 - 110304023 on Build GRCh38 | Gain | TXNDC8 AKAP2 TXN PALM2-AKAP2 C9orf152 |
esv2195369 | Chr.9:110067755 - 110068737 on Build GRCh38 | Deletion | AKAP2 PALM2-AKAP2 |
esv2501710 | Chr.9:110067316 - 110068996 on Build GRCh38 | Deletion | AKAP2 PALM2-AKAP2 |
nsv471314 | Chr.9:110059903 - 110093364 on Build GRCh38 | Loss | AKAP2 PALM2-AKAP2 |