Assay Details
Target Gene Details
Entrez Gene ID: | 27434 |
Gene Name: | DNA polymerase mu |
Gene Aliases: |
Pol Mu, Tdt-N |
Location: |
Chr.7:44072062-44082540 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830983 | Chr.7:44068842 - 44201624 on Build GRCh38 | Loss | POLD2 GCK AEBP1 MIR4649 YKT6 MIR6838 POLM MYL7 |
nsv8082 | Chr.7:43956156 - 44207229 on Build GRCh38 | Gain | POLD2 LINC00957 GCK MIR6837 YKT6 RASA4CP PGAM2 MYL7 DBNL POLR2J4 AEBP1 MIR4649 MIR6838 SPDYE1 POLM |
nsv428165 | Chr.7:43855828 - 44172960 on Build GRCh38 | Gain | URGCP POLD2 LINC00957 GCK MIR6837 RASA4CP PGAM2 MRPS24 MYL7 LOC107986791 DBNL POLR2J4 AEBP1 MIR4649 MIR6838 URGCP-MRPS24 SPDYE1 POLM UBE2D4 |
nsv470210 | Chr.7:44045936 - 44139229 on Build GRCh38 | Loss | POLD2 DBNL MIR6837 AEBP1 MIR4649 MIR6838 PGAM2 POLM MYL7 |
nsv830980 | Chr.7:43865998 - 44078539 on Build GRCh38 | Loss | URGCP LINC00957 MIR6837 RASA4CP PGAM2 MRPS24 LOC107986791 DBNL POLR2J4 MIR6838 URGCP-MRPS24 SPDYE1 POLM UBE2D4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |