Assay Details
Target Gene Details
Entrez Gene ID: | 27434 |
Gene Name: | DNA polymerase mu |
Gene Aliases: |
Pol Mu, Tdt-N |
Location: |
Chr.7:44072062-44082540 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830983 | Chr.7:44068842 - 44201624 on Build GRCh38 | Loss | MIR6838 YKT6 POLD2 MYL7 POLM MIR4649 GCK AEBP1 |
nsv8082 | Chr.7:43956156 - 44207229 on Build GRCh38 | Gain | MIR6838 RASA4CP LINC00957 YKT6 POLD2 POLM MIR4649 POLR2J4 GCK SPDYE1 AEBP1 DBNL MYL7 MIR6837 PGAM2 |
nsv428165 | Chr.7:43855828 - 44172960 on Build GRCh38 | Gain | MIR6838 RASA4CP LINC00957 POLD2 URGCP POLM MIR4649 POLR2J4 GCK SPDYE1 LOC107986791 AEBP1 MRPS24 DBNL URGCP-MRPS24 MYL7 MIR6837 PGAM2 UBE2D4 |
nsv470210 | Chr.7:44045936 - 44139229 on Build GRCh38 | Loss | DBNL MIR6838 POLD2 MYL7 MIR6837 POLM MIR4649 PGAM2 AEBP1 |
nsv830980 | Chr.7:43865998 - 44078539 on Build GRCh38 | Loss | MIR6838 RASA4CP LINC00957 URGCP POLM POLR2J4 SPDYE1 LOC107986791 MRPS24 DBNL URGCP-MRPS24 MIR6837 PGAM2 UBE2D4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |