Assay Details
Target Gene Details
Entrez Gene ID: | 1551 |
Gene Name: | cytochrome P450 family 3 subfamily A member 7 |
Gene Aliases: |
CP37, CYPIIIA7, P-450(HFL33), P-450111A7, P450-HFLA |
Location: |
Chr.7:99705037-99735200 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYP3A7 | NM_000765.4 | NP_000756.3 | ||
BC067436.1 | AAH67436.1 | |||
BX647143.1 | ||||
D00408.1 | BAA00310.1 |
Target Gene Details
Entrez Gene ID: | 100861540 |
Gene Name: | CYP3A7-CYP3A51P readthrough |
Gene Aliases: |
CYP3A7-3AP1, CYP3A7-CYP3AP1, CYP3A7.1L |
Location: |
Chr.7:99684679-99735200 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYP3A7-CYP3A51P | NM_001256497.2 | NP_001243426.2 | ||
AF315325.1 | AAG48618.1 |
Target Gene Details
Entrez Gene ID: | 221785 |
Gene Name: | zinc finger and SCAN domain containing 25 |
Gene Aliases: |
ZNF498 |
Location: |
Chr.7:99616600-99738689 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZSCAN25 | BX640720.1 | CAE45839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734887 | Chr.7:99697899 - 99750491 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv522107 | Chr.7:99703135 - 99717363 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv970911 | Chr.7:99677869 - 99735967 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)