Assay Details
Target Gene Details
Entrez Gene ID: | 3569 |
Gene Name: | interleukin 6 |
Gene Aliases: |
BSF-2, BSF2, CDF, HGF, HSF, IFN-beta-2, IFNB2, IL-6 |
Location: |
Chr.7:22725889-22732002 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL6 | NM_000600.4 | NP_000591.1 | ||
NM_001318095.1 | NP_001305024.1 | |||
XM_005249745.4 | 3 | 1056 | XP_005249802.1 | |
XM_011515390.2 | XP_011513692.1 | |||
A09363.1 | CAA00839.1 | |||
AK298013.1 | 3 | 1020 | ||
AK298077.1 | 3 | 720 | ||
AK301141.1 | 4 | 911 | ||
BC015511.1 | AAH15511.1 | |||
BT019748.1 | AAV38553.1 | |||
BT019749.1 | AAV38554.1 | |||
CD013918.1 | ||||
CR450296.1 | CAG29292.1 | |||
M14584.1 | AAA52728.1 | |||
M18403.1 | AAA52729.1 | |||
M29150.1 | AAA59154.1 | |||
M54894.1 | AAC41704.1 | |||
S56892.1 | AAD13886.1 | |||
X04403.1 | CAA27991.1 | |||
X04430.1 | CAA28026.1 | |||
X04602.1 | CAA28268.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759518 | Chr.7:22715965 - 22881291 on Build GRCh38 | Loss | SNORD93 TOMM7 LOC541472 IL6 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |