Assay Details
Target Gene Details
Entrez Gene ID: | 51547 |
Gene Name: | sirtuin 7 |
Gene Aliases: |
SIR2L7 |
Location: |
Chr.17:81911939-81918182 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIRT7 | NM_016538.2 | NP_057622.1 | ||
XM_006722284.2 | XP_006722347.1 | |||
XM_011523580.1 | XP_011521882.1 | |||
XM_011523581.2 | XP_011521883.1 | |||
AF233395.1 | AAF43431.1 | |||
AK002027.1 | 5 | 1043 | BAA92044.1 | |
AK094326.1 | ||||
AK131437.1 | ||||
AK131484.1 | ||||
AK290265.1 | ||||
AL137626.1 | CAB70848.2 | |||
BC017305.2 | AAH17305.1 | |||
BC101791.1 | AAI01792.1 | |||
BC101793.1 | AAI01794.1 | |||
BQ890203.1 | ||||
CD579273.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv29964 | Chr.17:81891311 - 81952513 on Build GRCh38 | Loss | SIRT7 PYCR1 NOTUM MAFG-AS1 MYADML2 ANAPC11 ALYREF PCYT2 MAFG NPB |
nsv833571 | Chr.17:81913391 - 81979943 on Build GRCh38 | Loss | ASPSCR1 SIRT7 PYCR1 NOTUM MAFG-AS1 MYADML2 MAFG |
nsv952392 | Chr.17:81885825 - 82065724 on Build GRCh38 | Deletion | SIRT7 PYCR1 NOTUM MYADML2 DUS1L ANAPC11 PCYT2 RFNG GPS1 STRA13 NPB ASPSCR1 DCXR MAFG-AS1 RAC3 ALYREF MAFG LRRC45 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | WDR45B SIRT7 NOTUM CEP131 FAAP100 RAB40B GCGR RAC3 LOC105376791 SLC38A10 CCDC137 CCDC57 AATK-AS1 CSNK1D ACTG1 ZNF750 MYADML2 TEX19 DUS1L LINC00482 PCYT2 FN3K GPS1 STRA13 NPB ARL16 ASPSCR1 CD7 BAHCC1 TBCD FOXK2 SNORD134 MIR3186 UTS2R ALYREF PDE6G LOC101929552 B3GNTL1 SECTM1 LOC105376790 NARF OXLD1 LOC105371925 TSPAN10 FSCN2 LOC100130370 MCRIP1 RFNG ARHGDIA MIR4525 MIR6786 SLC16A3 TEPSIN DCXR MAFG-AS1 SLC25A10 PPP1R27 OGFOD3 HGS MRPL12 LRRC45 PYCR1 MIR4740 FN3KRP MIR6787 C17orf89 P4HB ANAPC11 AATK TMEM105 FASN LOC101929511 C17orf62 NPLOC4 HEXDC MAFG LOC105371944 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | SIRT7 NOTUM OXLD1 TSPAN10 FAAP100 FSCN2 MCRIP1 RFNG ARHGDIA MIR6786 GCGR DCXR MAFG-AS1 RAC3 SLC25A10 PPP1R27 CCDC137 HGS MRPL12 LRRC45 PYCR1 P4HB MYADML2 DUS1L ANAPC11 PCYT2 GPS1 STRA13 NPB ARL16 ASPSCR1 NPLOC4 ALYREF PDE6G MAFG |
esv33468 | Chr.17:81891535 - 81927756 on Build GRCh38 | Loss | SIRT7 ANAPC11 ALYREF PCYT2 MAFG NPB |
nsv833569 | Chr.17:81838937 - 82011412 on Build GRCh38 | Loss | SIRT7 PYCR1 NOTUM P4HB MYADML2 ANAPC11 PCYT2 ARHGDIA NPB ASPSCR1 MAFG-AS1 ALYREF MAFG |
esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss | WDR45B SIRT7 NOTUM RAB40B RAC3 LOC105376791 CCDC57 CSNK1D ZNF750 MYADML2 TEX19 DUS1L PCYT2 FN3K GPS1 STRA13 NPB ASPSCR1 CD7 TBCD FOXK2 SNORD134 UTS2R ALYREF LOC101929552 B3GNTL1 SECTM1 LOC105376790 NARF MCRIP1 RFNG ARHGDIA MIR4525 SLC16A3 DCXR MAFG-AS1 PPP1R27 OGFOD3 LRRC45 METRNL PYCR1 FN3KRP MIR6787 P4HB ANAPC11 FASN LOC101929511 C17orf62 HEXDC MAFG LOC105371944 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |