Assay Details
Target Gene Details
Entrez Gene ID: | 51547 |
Gene Name: | sirtuin 7 |
Gene Aliases: |
SIR2L7 |
Location: |
Chr.17:81911939-81918182 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIRT7 | NM_016538.2 | 10 | 1137 | NP_057622.1 |
XM_006722284.2 | 8 | 871 | XP_006722347.1 | |
XM_011523580.1 | 9 | 1003 | XP_011521882.1 | |
XM_011523581.2 | 9 | 2740 | XP_011521883.1 | |
AF233395.1 | 10 | 1119 | AAF43431.1 | |
AK002027.1 | 8 | 1917 | BAA92044.1 | |
AK094326.1 | 10 | 1289 | ||
AK131437.1 | 6 | 2320 | ||
AK131484.1 | 4 | 2851 | ||
AK290265.1 | 10 | 1105 | ||
AL137626.1 | 7 | 705 | CAB70848.2 | |
BC017305.2 | 10 | 1101 | AAH17305.1 | |
BC101791.1 | 10 | 1118 | AAI01792.1 | |
BC101793.1 | 10 | 1117 | AAI01794.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv29964 | Chr.17:81891311 - 81952513 on Build GRCh38 | Loss | NOTUM ALYREF MYADML2 PCYT2 ANAPC11 NPB MAFG-AS1 MAFG SIRT7 PYCR1 |
nsv952392 | Chr.17:81885825 - 82065724 on Build GRCh38 | Deletion | ALYREF ASPSCR1 RAC3 DUS1L RFNG PCYT2 ANAPC11 PYCR1 NOTUM MYADML2 STRA13 NPB LRRC45 DCXR GPS1 MAFG-AS1 MAFG SIRT7 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | SLC25A10 ALYREF FOXK2 LOC105376791 DUS1L RFNG RAB40B ACTG1 TSPAN10 PDE6G PYCR1 FN3KRP FN3K P4HB CEP131 SLC16A3 LOC101929552 LINC00482 ARL16 HEXDC CCDC57 B3GNTL1 SIRT7 C17orf62 C17orf89 RAC3 GCGR TMEM105 CSNK1D BAHCC1 FASN LOC100130370 MIR6786 LOC101929511 LRRC45 AATK-AS1 MIR4525 HGS MIR3186 SNORD134 LOC105371944 AATK OGFOD3 ZNF750 ANAPC11 LOC105371925 NOTUM NPLOC4 NARF WDR45B STRA13 TEX19 NPB MAFG ASPSCR1 MRPL12 UTS2R MIR6787 MCRIP1 PCYT2 CCDC137 FSCN2 TBCD SECTM1 LOC105376790 TEPSIN SLC38A10 FAAP100 MYADML2 MIR4740 OXLD1 ARHGDIA CD7 DCXR GPS1 MAFG-AS1 PPP1R27 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | SLC25A10 ALYREF HGS DUS1L RFNG ANAPC11 TSPAN10 PDE6G PYCR1 NOTUM NPLOC4 P4HB STRA13 ARL16 NPB MAFG SIRT7 ASPSCR1 MRPL12 RAC3 MCRIP1 PCYT2 GCGR CCDC137 FSCN2 MIR6786 FAAP100 MYADML2 OXLD1 ARHGDIA LRRC45 DCXR GPS1 MAFG-AS1 PPP1R27 |
esv33468 | Chr.17:81891535 - 81927756 on Build GRCh38 | Loss | ALYREF PCYT2 ANAPC11 NPB MAFG SIRT7 |
nsv833569 | Chr.17:81838937 - 82011412 on Build GRCh38 | Loss | ALYREF ASPSCR1 PCYT2 ANAPC11 PYCR1 NOTUM P4HB MYADML2 ARHGDIA NPB MAFG-AS1 MAFG SIRT7 |
esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss | ALYREF METRNL FOXK2 LOC105376791 DUS1L RFNG RAB40B PYCR1 FN3KRP FN3K P4HB SLC16A3 LOC101929552 HEXDC CCDC57 B3GNTL1 SIRT7 C17orf62 RAC3 CSNK1D FASN LOC101929511 LRRC45 MIR4525 SNORD134 LOC105371944 OGFOD3 ZNF750 ANAPC11 NOTUM NARF WDR45B STRA13 TEX19 NPB MAFG ASPSCR1 UTS2R MIR6787 MCRIP1 PCYT2 TBCD SECTM1 LOC105376790 MYADML2 ARHGDIA CD7 DCXR GPS1 MAFG-AS1 PPP1R27 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |