Assay Details
Target Gene Details
Entrez Gene ID: | 10865 |
Gene Name: | AT-rich interaction domain 5A |
Gene Aliases: |
MRF-1, MRF1, RP11-363D14 |
Location: |
Chr.2:96536719-96552638 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARID5A | NM_001319085.1 | NP_001306014.1 | ||
NM_001319087.1 | NP_001306016.1 | |||
NM_001319092.1 | 1 | 388 | NP_001306021.1 | |
NM_001319093.1 | NP_001306022.1 | |||
NM_001319094.1 | NP_001306023.1 | |||
NM_001319096.1 | NP_001306025.1 | |||
NM_212481.2 | NP_997646.1 | |||
XM_017003183.1 | 1 | 257 | XP_016858672.1 | |
XM_017003184.1 | XP_016858673.1 | |||
XM_017003185.1 | 1 | 254 | XP_016858674.1 | |
AK300173.1 | ||||
AK304566.1 | ||||
AK307768.1 | ||||
BM556343.1 | ||||
BX396215.2 | ||||
BX648918.1 | ||||
CR981009.1 | ||||
HY002073.1 | ||||
HY076710.1 | ||||
M62324.1 | AAA36325.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2720413 | Chr.2:96472591 - 96793459 on Build GRCh38 | Deletion | FER1L5 ARID5A LMAN2L NEURL3 CNNM4 LOC105373496 KANSL3 |
dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion | STARD7-AS1 DUSP2 NEURL3 ANKRD39 ANKRD36B SNRNP200 LOC105373496 LOC100506076 SEMA4C NCAPH CNNM4 LOC101927053 ASTL CIAO1 LOC105373495 ANKRD36 TMEM127 FAM178B ARID5A LOC107984110 STARD7 LMAN2L ITPRIPL1 FAHD2CP CNNM3 FER1L5 MIR3127 ANKRD23 FAHD2B ANKRD36C GPAT2 KANSL3 ADRA2B LOC100506123 LOC653924 |
nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion | STARD7-AS1 DUSP2 NEURL3 ANKRD39 ANKRD36B SNRNP200 LOC105373496 LOC100506076 SEMA4C NCAPH CNNM4 LOC101927053 ASTL CIAO1 LOC105373495 ANKRD36 TMEM127 FAM178B ARID5A LOC107984110 STARD7 LMAN2L ITPRIPL1 FAHD2CP CNNM3 FER1L5 MIR3127 ANKRD23 FAHD2B ANKRD36C GPAT2 KANSL3 ADRA2B LOC100506123 LOC653924 |
esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication | STARD7-AS1 DUSP2 NEURL3 ANKRD39 ANKRD36B SNRNP200 LOC105373496 LOC100506076 SEMA4C NCAPH CNNM4 LOC101927053 ASTL CIAO1 LOC105373495 ANKRD36 TMEM127 FAM178B ARID5A LOC107984110 STARD7 LMAN2L ITPRIPL1 FAHD2CP CNNM3 FER1L5 MIR3127 ANKRD23 FAHD2B ANKRD36C GPAT2 KANSL3 ADRA2B LOC100506123 LOC653924 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |