Assay Details
Target Gene Details
Entrez Gene ID: | 57628 |
Gene Name: | dipeptidyl peptidase like 10 |
Gene Aliases: |
DPL2, DPPY, DPRP-3, DPRP3 |
Location: |
Chr.2:114442358-115845752 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPP10 | NM_001178036.1 | NP_001171507.1 | ||
NM_001178037.1 | NP_001171508.1 | |||
NM_001321905.1 | NP_001308834.1 | |||
NM_001321907.1 | NP_001308836.1 | |||
NM_001321908.1 | NP_001308837.1 | |||
NM_001321910.1 | NP_001308839.1 | |||
NM_001321911.1 | NP_001308840.1 | |||
NM_001321912.1 | NP_001308841.1 | |||
NM_001321913.1 | NP_001308842.1 | |||
NM_020868.4 | NP_065919.2 | |||
AK055256.1 | ||||
AK289967.1 | ||||
AK315147.1 | ||||
AY172661.3 | AAO17263.2 | |||
BC030832.1 | AAH30832.1 | |||
DQ857321.1 |
Target Gene Details
Entrez Gene ID: | 389023 |
Gene Name: | DPP10 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:115144048-115161343 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPP10-AS1 | NR_036580.1 | |||
AB593078.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893137 | Chr.2:114489425 - 116378222 on Build GRCh38 | Gain | DPP10-AS3 DPP10-AS1 DPP10 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |