Assay Details
Target Gene Details
Entrez Gene ID: | 123624 |
Gene Name: | ATP/GTP binding protein like 1 |
Gene Aliases: |
CCP4, FECD8 |
Location: |
Chr.15:86079635-87031476 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AGBL1 | XM_011521227.2 | XP_011519529.1 | ||
DA695799.1 |
Target Gene Details
Entrez Gene ID: | 102724452 |
Gene Name: | uncharacterized LOC102724452 |
Gene Aliases: |
- |
Location: |
Chr.15:86938797-86988426 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC102724452 | NR_135683.1 | |||
BX100197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1039254 | Chr.15:86527126 - 87399757 on Build GRCh38 | Loss | LOC102724452 AGBL1 LOC105370954 |
nsv519797 | Chr.15:86084875 - 87238541 on Build GRCh38 | Gain | LOC102724452 LINC01584 AGBL1-AS1 AGBL1 LOC105370954 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |