Assay Details
Target Gene Details
Entrez Gene ID: | 55068 |
Gene Name: | ecto-NOX disulfide-thiol exchanger 1 |
Gene Aliases: |
CNOX, PIG38, bA64J21.1, cCNOX |
Location: |
Chr.13:43213525-43786980 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENOX1 | NM_001127615.1 | NP_001121087.1 | ||
NM_001242863.1 | NP_001229792.1 | |||
NM_017993.3 | NP_060463.2 | |||
XM_005266439.3 | XP_005266496.1 | |||
XM_011535126.2 | XP_011533428.1 | |||
XM_011535127.2 | XP_011533429.1 | |||
XM_011535128.2 | XP_011533430.1 | |||
XM_011535132.2 | XP_011533434.1 | |||
XM_017020637.1 | XP_016876126.1 | |||
XM_017020638.1 | XP_016876127.1 | |||
XM_017020639.1 | XP_016876128.1 | |||
XM_017020640.1 | XP_016876129.1 | |||
XM_017020641.1 | XP_016876130.1 | |||
XM_017020642.1 | XP_016876131.1 | |||
AK000956.1 | BAA91442.1 | |||
BC024178.1 | AAH24178.1 | |||
BM834061.1 | ||||
DA512678.1 | ||||
DA779748.1 | ||||
DB058776.1 |
Target Gene Details
Entrez Gene ID: | 100874130 |
Gene Name: | ENOX1 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.13:43448852-43459488 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENOX1-AS2 | NR_120399.1 | |||
CR739820.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892336 | Chr.13:43420155 - 43603319 on Build GRCh38 | Loss | ENOX1 ENOX1-AS2 |
nsv516714 | Chr.13:43450021 - 43454653 on Build GRCh38 | Loss | ENOX1 ENOX1-AS2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |