Assay Details
Target Gene Details
Entrez Gene ID: | 79903 |
Gene Name: | N(alpha)-acetyltransferase 60, NatF catalytic subunit |
Gene Aliases: |
HAT4, NAT15 |
Location: |
Chr.16:3443611-3486963 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAA60 | NM_001083601.2 | NP_001077070.1 | ||
NM_001317093.1 | NP_001304022.1 | |||
AK297219.1 | ||||
AL833594.1 | ||||
DA786716.1 | ||||
DC401343.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv29492 | Chr.16:3442826 - 3448744 on Build GRCh38 | Gain | NAA60 ZNF597 |
esv3637667 | Chr.16:3442554 - 3448680 on Build GRCh38 | Gain | NAA60 ZNF597 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |